missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lurasidone Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Lurasidone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Lurasidone HCl |
| CAS | 367514-88-3 |
| Analyte or Component Names | Lurasidone HCl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H36 N4 O2 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4,7-Methano-1H-isoindole-1,3(2H)-dione, 2-[[(1R,2R)-2-[[4-(1,2-benzisothiazol-3-yl)-1-piperazinyl]methyl]cyclohexyl]methyl]hexahydro-, hydrochloride (1:1), (3aR,4S,7R,7aS)-, 4,7-Methano-1H-isoindole-1,3(2H)-dione, 2-[[(1R,2R)-2-[[4-(1,2-benzisothiazol-3-yl)-1-piperazinyl]methyl]cyclohexyl]methyl]hexahydro-, monohydrochloride, (3aR,4S,7R,7aS)- (9CI), Latuda, Lurasidone hydrochloride, SM 13496 |
| InChI Formula | InChI=1S/C28H36N4O2S.ClH/c33-27-24-18-9-10-19(15-18)25(24)28(34)32(27)17-21-6-2-1-5-20(21)16-30-11-13-31(14-12-30)26-22-7-3-4-8-23(22)35-29-26;/h3-4,7-8,18-21,24-25H,1-2,5-6,9-17H2;1H/t18-,19+,20-,21-,24+,25-;/m0./s1 |
| SMILES | Cl.O=C1[C@@H]2[C@H]3CC[C@H](C3)[C@@H]2C(=O)N1C[C@@H]4CCCC[C@H]4CN5CCN(CC5)c6nsc7ccccc67 |
| Molecular Weight (g/mol) | 529.14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction