missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G6F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24668SS
This item is not returnable.
View return policy
Description
Ly-6G6F Polyclonal specifically detects Ly-6G6F in Human, Goat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ly-6G6F | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| C6orf21, G6F, locus G6D, LY6G6D, lymphocyte antigen 6 complex locus protein G6f, lymphocyte antigen 6 complex, locus G6F, NG32 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, horse, rat, rhesus monkey and dog, 90% homologous in cow, 80% homologous in platypus and 70% homologous in opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 1.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Q5SQ64 | |
| LY6G6F | |
| A synthetic peptide corresponding to amino acids 66-100 of human G6D was used as immunogen, GenBank no NP_001003693. | |
| 0.025 mg | |
| Signal Transduction | |
| 259215 | |
| Human, Goat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction