missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G6F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00 € - 372.00 €
Specifications
| Antigen | Ly-6G6F |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18168581
|
Novus Biologicals
NBP2-24668SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18735243
|
Novus Biologicals
NBP2-24668 |
372.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ly-6G6F Polyclonal specifically detects Ly-6G6F in Human, Goat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Ly-6G6F | |
| Western Blot 1.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Goat, Primate | |
| Q5SQ64 | |
| 259215 | |
| A synthetic peptide corresponding to amino acids 66-100 of human G6D was used as immunogen, GenBank no NP_001003693. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| C6orf21, G6F, locus G6D, LY6G6D, lymphocyte antigen 6 complex locus protein G6f, lymphocyte antigen 6 complex, locus G6F, NG32 | |
| LY6G6F | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, horse, rat, rhesus monkey and dog, 90% homologous in cow, 80% homologous in platypus and 70% homologous in opossum. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title