Learn More
Ly-6G6F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-24668
 View more versions of this product
Product Code. 18735243
  • 372.00 € / 0.10mg
Description
Ly-6G6F Polyclonal specifically detects Ly-6G6F in Human, Goat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Purified