missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6K Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24405SS
This item is not returnable.
View return policy
Description
Ly6K Polyclonal specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Ly6K | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LY6K | |
| Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein. | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot, ELISA 1:100-1:2000 | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54742 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction