missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6K Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
225.00 € - 386.00 €
Specifications
| Antigen | Ly6K |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18135484
|
Novus Biologicals
NBP2-24405SS |
0.025 mg |
225.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710514
|
Novus Biologicals
NBP2-24405 |
386.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ly6K Polyclonal specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA.Specifications
| Ly6K | |
| Western Blot, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| LY6K | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 54742 | |
| Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title