missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin-alpha/TNF-beta, Mouse anti-Human, Clone: 3F12.2D3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-81106-0.025MG
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Flow Cytometry, ELISA, Surface Plasmon Resonance, Neutralization.
Specifications
| Lymphotoxin-alpha/TNF-beta | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Surface Plasmon Resonance, Neutralization | |
| LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| 3F12 binds to human LTalpha3 and LT-alpha-beta (with a KD of ∽0.3 nM, determined by BIACORE; ∽ 37 pM, determined by ELISA) in both its soluble homotrimeric and membrane heterotrimeric forms. LTalpha belongs to the tumor necrosis factor (TNF) superfamily and is secreted as a homo-trimer (LTalpha3), or is expressed on the cell surface in complex with LTbeta. Lymphotoxin is produced by lymphocytes and in its homotrimeric form binds to TNFRSF1A/TNFR1, TNFRSF1B/TNFBR and TNFRSF14/HVEM. LTalpha3 signaling induces target cells to upregulate many chemokines and cytokines in an NFkB-dependent manner. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Neutralization | |
| 3F12.2D3 | |
| Unconjugated | |
| PBS | |
| LTA | |
| 3F12 was prepared by hyperimmunizing BALB/c mice with purified recombinant human LTalpha expressed in E. coli - the hybridoma clone was selected for its ability to bind LTalpha3 by ELISA. The CDRs of the antibody were grafted to the framework regions of the mice mAb 2G7 to create the chimeric version of the antibody. | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 4049 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction