missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NBP1-49600
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/ml | |
| Western Blot 1:10000, Simple Western 1:12.5, Chromatin Immunoprecipitation reported in scientific literature (PMID 23884959; 23129632), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23129632), Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) | |
| Q9H3R0, Q8VCD7 | |
| KDM4C | |
| A partial recombinant protein made to an internal region of the human JMJD2C protein (within residues 450-600). [Swiss-Prot Q9H3R0] | |
| 0.1 mL | |
| Chromatin Modifiers, Epigenetics | |
| 23081 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction