missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
244.00 € - 527.00 €
Specifications
| Antigen | Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:10000, Simple Western 1:12.5, Chromatin Immunoprecipitation reported in scientific literature (PMID 23884959; 23129632), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23129632), Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058005
|
Novus Biologicals
NBP1-49600SS |
0.025 mL |
244.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745693
|
Novus Biologicals
NBP1-49600 |
0.1 mL |
527.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C | |
| Western Blot 1:10000, Simple Western 1:12.5, Chromatin Immunoprecipitation reported in scientific literature (PMID 23884959; 23129632), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23129632), Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Epigenetics | |
| PBS with 0.05% Sodium Azide | |
| EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, lysine-specific demethylase 4C | |
| KDM4C | |
| IgG | |
| Affinity Purified |
| 1.0 mg/ml | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9H3R0, Q8VCD7 | |
| 23081 | |
| A partial recombinant protein made to an internal region of the human JMJD2C protein (within residues 450-600). [Swiss-Prot Q9H3R0] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title