missing translation for 'onlineSavingsMsg'
Learn More

m-Acetotoluidide, Thermo Scientific™

Product Code. p-3593451 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10493925

Brand: Thermo Scientific Acros 391300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 537-92-8
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00014962
InChI Key ALMHSXDYCFOZQD-UHFFFAOYSA-N
Synonym 3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
PubChem CID 10843
IUPAC Name N-(3-methylphenyl)acetamide
SMILES CC1=CC(=CC=C1)NC(=O)C

Specifications

Melting Point 65°C to 67°C
Boiling Point 303°C
Packaging Glass bottle
Linear Formula CH3C6H4NHC(O)CH3
Quantity 5 g
Merck Index 15, 72
Formula Weight 149.19
Chemical Name or Material m-Acetotoluidide
compliance-icons
Product Identifier
  • m-Acetotoluidide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.