missing translation for 'onlineSavingsMsg'
Learn More

Macro H2A.2 Antibody (CL5263) [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30530162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30530162 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30530162 Supplier Novus Biologicals Supplier No. NBP344246AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Macro H2A.2 Monoclonal antibody specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macro H2A.2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL5263
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q9P0M6, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55506
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.