missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MafA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24636
This item is not returnable.
View return policy
Description
MafA Polyclonal specifically detects MafA in Human, Mouse, Rat, Opossum samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP).
Specifications
| MafA | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian) | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and bovine, 87% homologous in pufferfish and 81% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.25-0.5ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 24509877), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24509877), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24509877), ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP) | |
| Q8NHW3 | |
| MAFA | |
| A portion of amino acids 300-350 of human MafA was used as the immunogen. | |
| 0.1 mg | |
| Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 389692 | |
| Human, Mouse, Rat, Opossum | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction