missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MafA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
226.00 € - 372.00 €
Specifications
| Antigen | MafA |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.25-0.5ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 24509877), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24509877), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24509877), ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18156461
|
Novus Biologicals
NBP2-24636SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799833
|
Novus Biologicals
NBP2-24636 |
0.1 mg |
372.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MafA Polyclonal specifically detects MafA in Human, Mouse, Rat, Opossum samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP).Specifications
| MafA | |
| Western Blot 0.25-0.5ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 24509877), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24509877), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24509877), ICC/IF (Negative), Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Opossum | |
| Q8NHW3 | |
| 389692 | |
| A portion of amino acids 300-350 of human MafA was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian) | |
| MAFA | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, pig and bovine, 87% homologous in pufferfish and 81% homologous in zebrafish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title