missing translation for 'onlineSavingsMsg'
Learn More

MAGEF1 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30565970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30565970 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30565970 Supplier Novus Biologicals Supplier No. NBP345969R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAGEF1 Polyclonal antibody specifically detects MAGEF1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEF1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias MAGE-F1 antigen, melanoma antigen family F, 1, melanoma-associated antigen F1, MGC19617
Host Species Rabbit
Immunogen melanoma antigen family F, 1. (Uniprot# Q9HAY2)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64110
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.