missing translation for 'onlineSavingsMsg'
Learn More

Magnesium salicylate tetrahydrate, tech.

Product Code. p-7050742
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 11308158

Brand: Thermo Scientific Alfa Aesar 039379.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18917-95-8 | C14H18MgO10 | 370.59 g/mol

It is used to treat mild to moderate muscular pain. It is also used to treat headaches, general back pain, and certain joint pains like arthritis. It is found in a variety of over-the-counter (OTC) medications as an anti-inflammatory, primarily for back-pain relief.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18917-95-8
Molecular Formula C14H18MgO10
Molecular Weight (g/mol) 370.59
MDL Number MFCD01766176,MFCD00045815
InChI Key NBQBEWAYWAMLJJ-UHFFFAOYSA-L
Synonym magnesium salicylate
PubChem CID 131674114
IUPAC Name magnesium;2-hydroxybenzoic acid;2-oxidobenzoate;hydrate
SMILES O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O

Specifications

Odor Odorless
Quantity 100 g
Sensitivity Air Sensitive
Merck Index 14,5672
Solubility Information Soluble in water and alcohol
Formula Weight 370.60 (298.60 Anhydrous)
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium salicylate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.