missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Major allergen I polypeptide chain 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23611-50UG
This item is not returnable.
View return policy
Description
Major allergen I polypeptide chain 2 Polyclonal specifically detects Major allergen I polypeptide chain 2 in Human, Mouse, Rat, Feline, Primate samples. It is validated for Western Blot, ELISA.
Specifications
| Major allergen I polypeptide chain 2 | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000 | |
| P30440 | |
| CH2 | |
| Synthetic peptides unique to Fel D1 protein | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| AG4, allergen Cat-1, allergen FdI, allergen Fel d I-B, CH2, CH2 major allergen I, polypeptide chain 2, CH2 major allergen I, polypeptide chain 2 [ Felis catus (domestic cat) ], Fel d I, major allergen I, polypeptide chain 2 | |
| Rabbit | |
| 50 μg | |
| 677879 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction