missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Major allergen I polypeptide chain 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
256.00 € - 710.00 €
Specifications
| Antigen | Major allergen I polypeptide chain 2 |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18123542
|
Novus Biologicals
NBP2-23611-50UG |
50 μg |
438.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149271
|
Novus Biologicals
NBP2-23611 |
100 μg |
710.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162592
|
Novus Biologicals
NBP2-23611-0.025MG |
0.025 mg |
256.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Major allergen I polypeptide chain 2 Polyclonal specifically detects Major allergen I polypeptide chain 2 in Human, Mouse, Rat, Feline, Primate samples. It is validated for Western Blot, ELISA.Specifications
| Major allergen I polypeptide chain 2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| P30440 | |
| 677879 | |
| Synthetic peptides unique to Fel D1 protein | |
| Primary |
| Western Blot 1:500, ELISA 1:10000 | |
| Polyclonal | |
| Rabbit | |
| AG4, allergen Cat-1, allergen FdI, allergen Fel d I-B, CH2, CH2 major allergen I, polypeptide chain 2, CH2 major allergen I, polypeptide chain 2 [ Felis catus (domestic cat) ], Fel d I, major allergen I, polypeptide chain 2 | |
| CH2 | |
| IgG |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title