missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, Mikromol™

Product Code. 17981036
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981036 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981036 Supplier Mikromol Supplier No. MM1439.000250
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Maleic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic Acid
CAS 110-16-7
Analyte or Component Names Maleic Acid
Shipping Condition Room Temperature
Molecular Formula C4 H4 O4
Certifications/Compliance ISO 17034
Synonym (Z)-2-Butenedioic acid, Maleic acid, (2Z)-Butene-2-dioic acid, Maleinic acid, Malezid CM, Scotchbond Multipurpose Etchant, Toxilic acid, cis-1,2-Ethylenedicarboxylic acid, cis-2-Butenedioic acid, cis-Butenedioic acid, Aspartic Acid Impurity H, Magnesium Aspartate Dihydrate Impurity I, Malic Acid Impurity B, Magnesium Aspartate Impurity I
InChI Formula InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-
SMILES OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
Quantity 250 mg
Formula Weight 116.011 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.