missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, LoGiCal

Product Code. 30458896
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458896 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458896 Supplier LoGiCal Supplier No. LGCFOR0654.00
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Mannitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Mannitol
CAS 69-65-8
Analyte or Component Names D-Mannitol
Shipping Condition Room Temperature
Molecular Formula C6 H14 O6
Certifications/Compliance ISO 9001
Synonym Mannitol, d-Mannitol, D-Mannitol, Cordycepic acid (6 CI,7 CI), Mannitol, D- (8 CI), Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C, Pearlitol 50 C
InChI Formula InChI=1 S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12 H,1-2H2/t3-,4-,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2 R,3 R,4 R,5 R)-hexane-1,2,3,4,5,6-hexol
Industry Type Forensics
Molecular Weight (g/mol) 182.17
Quantity 10 mg
Formula Weight 182.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.