missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAO-A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38738-25ul
This item is not returnable.
View return policy
Description
MAO-A Polyclonal specifically detects MAO-A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MAO-A | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P21397 | |
| MAOA | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MENQEKASIAGHMFDVVVIGGG | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| amine oxidase [flavin-containing] A, EC 1.4.3, EC 1.4.3.4, MAO-A, monoamine oxidase A, Monoamine oxidase type A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4128 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction