missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAO-A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 559.00 €
Specifications
| Antigen | MAO-A |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18177877
|
Novus Biologicals
NBP2-38738 |
0.1 mL |
559.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649875
|
Novus Biologicals
NBP2-38738-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAO-A Polyclonal specifically detects MAO-A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MAO-A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P21397 | |
| 4128 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MENQEKASIAGHMFDVVVIGGG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| amine oxidase [flavin-containing] A, EC 1.4.3, EC 1.4.3.4, MAO-A, monoamine oxidase A, Monoamine oxidase type A | |
| MAOA | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title