missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline Hydrochloride, Mikromol™

Product Code. 18091810
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091810 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18091810 Supplier Mikromol Supplier No. MM0820.00
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Maprotiline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maprotiline Hydrochloride
CAS 10347-81-6
Analyte or Component Names Maprotiline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H23 N . Cl H
Certifications/Compliance ISO 17034
Synonym 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (1:1), 9,10-Ethanoanthracene-9(10H)-propanamine, N-methyl-, hydrochloride (9CI), 9,10-Ethanoanthracene-9(10H)-propylamine, N-methyl-, hydrochloride (7CI,8CI), 9,10-Dihydro-9-(3-methylaminopropyl)-9,10-ethanoanthracene hydrochloride, 9,10-Ethanoanthracene-9-propylamine, 9,10-dihydro-N-methyl-, hydrochloride, Ba 34276, CIBA 34,276Ba, CIBA 34276 hydrochloride, Deprilept, Ladiomil, Ludiomil, Maprotiline hydrochloride, Psymion, 9-[gamma-(Methylamino)propyl]-9,10-dihydro-9,10-ethanoanthracene hydrochloride
InChI Formula InChI=1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H
SMILES Cl.CNCCCC12CCC(c3ccccc13)c4ccccc24
IUPAC Name N-Methyl-9,10-ethanoanthracene-9(10H)-propanamine hydrochloride
Molecular Weight (g/mol) 313.86
Quantity 250 mg
Formula Weight 313.1597 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.