missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATH1 Antibody (SR1642), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21904-100ul
This item is not returnable.
View return policy
Description
MATH1 Monoclonal antibody specifically detects MATH1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MATH1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1642 | |
| Western Blot 1:500-1:1000 | |
| ATH1protein atonal homolog 1, atonal homolog 1 (Drosophila), BHLHA14, bHLHa14Math1, Class A basic helix-loop-helix protein 14, hATH1, Helix-loop-helix protein hATH-1, MATH-1 | |
| A synthesized peptide derived from human MATH1/HATH1 (Uniprot #: Q92858) | |
| 100 μg | |
| Cancer | |
| 474 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction