missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATH1 Antibody (SR1642), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | MATH1 |
|---|---|
| Clone | SR1642 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603454
|
Novus Biologicals
NBP3-21904-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696504
|
Novus Biologicals
NBP3-21904-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MATH1 Monoclonal antibody specifically detects MATH1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MATH1 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATH1protein atonal homolog 1, atonal homolog 1 (Drosophila), BHLHA14, bHLHa14Math1, Class A basic helix-loop-helix protein 14, hATH1, Helix-loop-helix protein hATH-1, MATH-1 | |
| A synthesized peptide derived from human MATH1/HATH1 (Uniprot #: Q92858) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1642 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 474 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title