missing translation for 'onlineSavingsMsg'
Learn More

mCherry Antibody (1C51), Novus Biologicals™

Product Code. 18728303 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18728303 25µL
18751724 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18728303 Supplier Novus Biologicals Supplier No. NBP196752SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 48 publications

mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Single Cell Western.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Purification Method Protein G purified
Target Species Mouse, Insect
Form Purified

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.