Learn More
MDM2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27883-1-AP-150UL
 View more versions of this product
Product Code. 16875885
  • 410.00 € / 150µL
Description
The MDM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MDM2. This antibody recognizes human antigen. The MDM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
MDM2
Polyclonal
Unconjugated
150 μL
4193
IgG
Western Blot, Flow Cytometry
367 μg/mL