missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | MDM2 |
|---|---|
| Concentration | 367 μg/mL |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16885885
|
Proteintech
27883-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16875885
|
Proteintech
27883-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MDM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MDM2. This antibody recognizes human antigen. The MDM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| MDM2 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Human | |
| IgG |
| 367 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 4193 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title