missing translation for 'onlineSavingsMsg'
Learn More

MED9 Antibody, R&D Systems™

Product Code. 18120484 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18120484 0.1 mL 0.1mL
18457391 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18120484 Supplier Novus Biologicals Supplier No. NBP231639
  • ONLINE EXCLUSIVE PRICE
    628.19 €
    590.10 € / 0.1mL
    Save 38.09 €
    6% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

MED9 Polyclonal specifically detects MED9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

TRUSTED_SUSTAINABILITY

Specifications

Antigen MED9
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9NWA0
Gene Alias FLJ10193mediator of RNA polymerase II transcription, subunit 9 homolog, MED25mediator of RNA polymerase II transcription subunit 9, mediator complex subunit 9MGC138234, mediator of RNA polymerase II transcription, subunit 9 homolog (S. cerevisiae), mediator subunit 25
Gene Symbols MED9
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MASAGVAAGRQAEDVLPPTSDQPLPDTKPLPPP
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55090
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.