missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MEIS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3F12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 |
| Formulation | PBS (pH 7.3) 1% BSA, 50% glycerol |
| Gene Alias | homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1(NP_002389) produced in E.coli. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?