missing translation for 'onlineSavingsMsg'
Learn More

MEK1, p Ser218, p Ser222 Antibody, Novus Biologicals™

Product Code. 18109301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
Unit Size:
0.05mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18109301 0.025 mg 25µg
18146581 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18109301 Supplier Novus Biologicals Supplier No. NBP224542SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEK1 Polyclonal specifically detects MEK1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK1 (p Ser218, p Ser222)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml
Formulation 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen A portion of amino acids 200-250, containing phospho serine residues at positions 218 and 222, of human MEK1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Phosphatase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5604
Test Specificity The amino acid sequence used as immunogen for this RXFP1 antibody is 100% homologous in human, chimp and orangutan, and 83% homologous in horse, bovine, dog and cat.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.