missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312976
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312976 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312976 Supplier Dr. Ehrenstorfer Supplier No. DREA14862500AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meloxicam
CAS 71125-38-7
Analyte or Component Names Meloxicam
Shipping Condition Room Temperature
Molecular Formula C14 H13 N3 O4 S2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2H-1,2-Benzothiazine-3-carboxamide, 4-hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-, 1,1-dioxide, Coxflam, Maxicam, Melonex, Meloxicam, Metacam, Mobec, Mobic, Mobicox, Movalis, Movatec, Revmoksikam, UH-AC 62XX, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
InChI Formula InChI=1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,18H,1-2H3,(H,15,16,19)
SMILES CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ncc(C)s3
IUPAC Name 4-hydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 351.4
Quantity 1 mL
Formula Weight 351.0347 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.