missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mepyramine Maleate EP Impurity A, Mikromol™
Description
Mepyramine Maleate EP Impurity A, Mikromol
Specifications
Specifications
| Chemical Name or Material | N-(4-methoxybenzyl)pyridin-2-amine |
| CAS | 52818-63-0 |
| Analyte or Component Names | N-(4-methoxybenzyl)pyridin-2-amine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-(4-Methoxybenzyl)pyridin-2-amine, Mepyramine Maleate Imp. A (EP), Mepyramine Imp. A (EP), N-(4-Methoxybenzyl)pyridin-2-amine, Mepyramine Maleate Impurity A, Mepyramine Impurity A |
| InChI Formula | InChI=1S/C13H14N2O/c1-16-12-7-5-11(6-8-12)10-15-13-4-2-3-9-14-13/h2-9H,10H2,1H3,(H,14,15) |
| Solution Type | Neat |
| SMILES | COc1ccc(CNc2ccccn2)cc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?