missing translation for 'onlineSavingsMsg'
Learn More

Mesoridazine.besylate, Lipomed

Product Code. 30458948
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458948 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458948 Supplier Lipomed Supplier No. LPMMES1880BS10
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Mesoridazine.besylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesoridazine Besilate
CAS 32672-69-8
Analyte or Component Names Mesoridazine Besilate
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O S2 . C6 H6 O3 S
Synonym Thioridazine Hydrochloride Imp. B (EP), Thioridazine Imp. B (EP), 10 H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, benzenesulfonate (1:1)10 H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, monobenzenesulfonate (9 CI), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10 H-phenothiazine (1:1), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)phenothiazine (1:1) (8 CI), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate), Imp. B (EP) as Besilate: 10-[2-[(2 RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10 H-phenothiazine Besilate (MesoridazineBesilate)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCCCC1CCN2c3ccccc3Sc4ccc(cc24)S(=O)C.OS(=O)(=O)c5ccccc5
IUPAC Name benzenesulfonic acid;10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 544.75
Quantity 10 mg
Formula Weight 544.1524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.