missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metacycline Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Methacycline Hydrochloride |
| CAS | 3963-95-9 |
| Analyte or Component Names | Methacycline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H22 N2 O8 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (4S,4aR,5S,5aR,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide hydrochloride, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, (4S,4aR,5S,5aR,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5α,5aα,12aα)]-, 6-Demethyl-6-deoxy-5-hydroxy-6-methylenetetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methylene-5-hydroxytetracycline hydrochloride, 6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride, 6-Methylene-5-hydroxytetracycline chlorohydrate, 6-Methylene-5-hydroxytetracycline hydrochloride, 6-Methyleneoxytetracycline hydrochloride, Adriamicina, Ciclobiotic, Germiciclin, Londomycin, Metacyclin hydrochloride, Metacycline hydrochloride, Metadomus, Methacycline hydrochloride, Methacycline monohydrochloride, Metilenbiotic, Optimycin, Physiomycine, Rindex, Doxycycline Monohydrate Imp. B (EP) as Hydrochloride, Doxycycline Hyclate Impurity B as Hydrochloride, Doxycycline Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C22H22N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-6,10,14-15,17,25,27-29,32H,1H2,2-3H3,(H2,23,31);1H/t10-,14-,15+,17+,22+;/m1./s1 |
| Solution Type | Neat |
| SMILES | Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@@H]3C(=C)c4cccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?