missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methamphetamine Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62754
This item is not returnable.
View return policy
Description
Methamphetamine Polyclonal specifically detects Methamphetamine in Non-species specific samples. It is validated for ELISA.
Specifications
| Methamphetamine | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Sheep | |
| Protein G purified | |
| RUO | |
| NB100-62754 is specific for methamphetamine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 5.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| METH | |
| Para-amino-metamphetamine conjugated to keyhole limpet hemocyanin | |
| 0.5 mL | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction