missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methamphetamine Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
212.00 € - 484.00 €
Specifications
| Antigen | Methamphetamine |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18092966
|
Novus Biologicals
NB100-62754-0.1ML |
0.1 mL |
212.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18005524
|
Novus Biologicals
NB100-62754 |
0.5 mL |
484.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methamphetamine Polyclonal specifically detects Methamphetamine in Non-species specific samples. It is validated for ELISA.Specifications
| Methamphetamine | |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.09% Sodium Azide | |
| Para-amino-metamphetamine conjugated to keyhole limpet hemocyanin | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| Sheep | |
| Non-species specific | |
| METH | |
| IgG | |
| Protein G purified | |
| NB100-62754 is specific for methamphetamine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title