missing translation for 'onlineSavingsMsg'
Learn More

Methimazol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321933
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321933 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321933 Supplier Dr. Ehrenstorfer Supplier No. DREV15020200AL100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methimazol
CAS 60-56-0
Analyte or Component Names Methimazol
Shipping Condition +20°C
Molecular Formula C4 H6 N2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1-Methyl-2-(methylsulphanyl)-1H-imidazole, Thiamazole Imp. C (EP), 1H-Imidazole, 1-methyl-2-(methylthio)- (9CI), Imidazole, 1-methyl-2-(methylthio)- (8CI), 1-Methyl-2-(methylthio)imidazole, 2-Methylthio-3-methylimidazole, Methimazole USP Related Compound C, Methimazole USP RC C, Thiamazole, Methimazole, 1,3-Dihydro-1-methyl-2H-imidazole-2-thione, 1-Methylimidazole-2-thiol, 1-Methyl-2-mercaptoimidazole, Mercazolyl, 2H-Imidazole-2-thione, 1,3-dihydro-1-methyl-, 4-Imidazoline-2-thione, 1-methyl- (6CI,7CI), Imidazole-2-thiol, 1-methyl- (8CI), 1-Methyl-1,3-dihydroimidazole-2-thione, 1-Methyl-1H-imidazole-2-thiol, 1-Methyl-2-mercapto-1H-imidazole, 1-Methyl-4-imidazoline-2-thione, 1-Methylimidazole-2(3H)-thione, 1-Methylimidazole-2-thione, 2-Mercapto-1-methyl-1H-imidazole, 2-Mercapto-1-methylimidazole, 2-Mercapto-N-methylimidazole, Basolan, Danantizol, Favistan, Frentirox, MMI, Mercaptazole, Mercazole, Metazolo, Methimazol, Methylmercaptoimidazole, Metizol, Metothyrin, Metothyrine, Metotirin, N-Methyl-2-mercaptoimidazole, N-Methylimidazolethiol, NSC 38608, Strumazol, Tapazole, Thacapzol, Thiamazol, Thycapzol, Thymidazol, Thymidazole, Thyrozol
InChI Formula InChI=1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)
SMILES CN1C=CNC1=S
IUPAC Name 3-methyl-1H-imidazole-2-thione
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 114.17
Quantity 5 mL
Formula Weight 114.0252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.