missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-acetamido-5-chlorobenzoate, 97%, Thermo Scientific™

Product Code. 11412858
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11412858

Brand: Thermo Scientific Alfa Aesar A16826.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20676-54-4
Molecular Formula C10H10ClNO3
Molecular Weight (g/mol) 227.64
MDL Number MFCD00144759
InChI Key TVAAIYFBEWHVCV-UHFFFAOYSA-N
Synonym 2-acetamido-5-chlorobenzoic acid methyl ester, methyl 5-chloro-2-acetamidobenzoate, methyl n-acetyl-5-chloroanthranilate, n-acetyl-5-chloroanthranilic acid methyl ester, 2-acetylamino-5-chloro-benzoic acid methyl ester, methyl 2-acetylamino-5-chlorobenzoate, pubchem3337, acmc-1cioq, methyl2-acetamido-5-chlorobenzoate, methyl 2-acetamido-5-chloranyl-benzoate
PubChem CID 4640571
IUPAC Name methyl 2-acetamido-5-chlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC=C1NC(C)=O

Specifications

Melting Point 128°C to 131°C
Quantity 50 g
Beilstein 2808490
Formula Weight 227.65
Percent Purity 97%
Chemical Name or Material Methyl 2-acetamido-5-chlorobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.