missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-chlorobenzoate, 98+%

Product Code. p-7048067
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11339016

Brand: Thermo Scientific Alfa Aesar L20187.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5202-89-1 | C8H8ClNO2 | 185.607 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5202-89-1
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00007837
InChI Key IGHVUURTQGBABT-UHFFFAOYSA-N
Synonym methyl 5-chloroanthranilate, methyl2-amino-5-chlorobenzoate, 5-chloroanthranilic acid methyl ester, benzoic acid, 2-amino-5-chloro-, methyl ester, 2-amino-5-chlorobenzoic acid methyl ester, anthranilic acid, 5-chloro-, methyl ester, 2-amino-5-chloro-benzoic acid methyl ester, pubchem3346, acmc-1axns, methyl-5-chloroanthranilate
PubChem CID 78878
IUPAC Name methyl 2-amino-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)N

Specifications

Melting Point 67°C to 70°C
Boiling Point 168°C to 170°C (1 mmHg)
Quantity 5 g
Beilstein 1072894
Formula Weight 185.61
Percent Purity ≥98%
Chemical Name or Material Methyl 2-amino-5-chlorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.