missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Aminobenzoate (Methyl Anthranilate), Mikromol™

Product Code. 17962936
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962936 Supplier Mikromol Supplier No. MM0142.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 2-Aminobenzoate (Methyl Anthranilate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anthranilic Acid Methyl Ester
CAS 134-20-3
Analyte or Component Names Anthranilic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym Methylanthranilate, Methyl 2-aminobenzoate, 2-Aminobenzoic acid methyl ester
InChI Formula InChI=1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3
SMILES COC(=O)c1ccccc1N
IUPAC Name methyl 2-aminobenzoate
Molecular Weight (g/mol) 151.16
Quantity 100 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.