missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-3-methylbenzoate, 97%

Product Code. p-7025703
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11439267

Brand: Thermo Scientific Alfa Aesar B21341.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23287-26-5 | C9H10O3 | 166.18 g/mol

Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

Solubility
Soluble in ethanol, fixed Oils, Propylene Glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23287-26-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00020051
InChI Key SUHLUMKZPUMAFP-UHFFFAOYSA-N
Synonym methyl 3-methylsalicylate, methyl o-cresotinate, benzoic acid, 2-hydroxy-3-methyl-, methyl ester, levegal pt, 2-hydroxy-3-methylbenzoic acid methyl ester, 2,3-cresotic acid, methyl ester, methyl-3-methylsalicylate, methyl 2-hydroxy-3-methyl-benzoate, methyl o-cresotate, 3-methylsalicylic acid methyl ester
PubChem CID 31835
IUPAC Name methyl 2-hydroxy-3-methylbenzoate
SMILES COC(=O)C1=CC=CC(C)=C1O

Specifications

Melting Point 28°C to 30°C
Boiling Point 234°C to 236°C
Quantity 2 g
Solubility Information Soluble in ethanol,fixed Oils,Propylene Glycol. Insoluble in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material Methyl 2-hydroxy-3-methylbenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.