missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Methoxybenzoate, Mikromol™
Description
Methyl 2-Methoxybenzoate
Specifications
Specifications
| Chemical Name or Material | Methyl 2-methoxybenzoate |
| CAS | 606-45-1 |
| Analyte or Component Names | Methyl 2-methoxybenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methyl 2-Methoxybenzoate, Benzoic acid, o-methoxy-, methyl ester (4CI), o-Anisic acid, methyl ester (6CI,7CI,8CI), 2-Methoxybenzoic acid methyl ester, Methyl 2-anisate, Methyl 2-methoxybenzoate, Methyl o-anisate, Methyl o-methoxybenzoate, NSC 406256, o-Methoxybenzoic acid methyl ester, Methyl Salicylate Impurity M |
| InChI Formula | InChI=1S/C9H10O3/c1-11-8-6-4-3-5-7(8)9(10)12-2/h3-6H,1-2H3 |
| SMILES | COC(=O)c1ccccc1OC |
| IUPAC Name | methyl 2-methoxybenzoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?