missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nitrobenzoate, 98+%

Product Code. 11486176
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
missing translation for 'unitSize'
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486176 25 g 25g
11496176 100 g 100g
11406186 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11486176 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B23009.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-27-9 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-27-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007136
InChI Key AOXPHVNMBPFOFS-UHFFFAOYSA-N
Synonym methyl o-nitrobenzoate, benzoic acid, 2-nitro-, methyl ester, 2-nitro-benzoic acid methyl ester, benzoic acid, o-nitro-, methyl ester, 2-nitrobenzoic acid methyl ester, methyl-2-nitrobenzoate, methyl o-nitro benzoate, methyl ortho-nitrobenzoate, acmc-209ml4, nitrobenzoic acid methyl ester
PubChem CID 69072
IUPAC Name methyl 2-nitrobenzoate
SMILES COC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -13°C
Density 1.285
Boiling Point 275°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 25 g
Beilstein 2050101
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.