missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dihydroxybenzoate, 97%

Product Code. 11367486
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367486 2 g 2g
11377486 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367486 Supplier Thermo Scientific Alfa Aesar Supplier No. L09552.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2150-43-8 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-43-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016437
InChI Key CUFLZUDASVUNOE-UHFFFAOYSA-N
Synonym methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
PubChem CID 287064
IUPAC Name methyl 3,4-dihydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 134°C to 138°C
Odor Odorless
Quantity 2 g
Beilstein 1637939
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dihydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.