missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-2-hydroxybenzoate, 98%

Product Code. p-7027978
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11466067

Brand: Thermo Scientific Alfa Aesar B25077.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21702-79-4 | C8H6Br2O3 | 309.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21702-79-4
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00017534
InChI Key KISQISIKTRRMOX-UHFFFAOYSA-N
PubChem CID 624217
IUPAC Name methyl 3,5-dibromo-2-hydroxybenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1O

Specifications

Melting Point 150°C to 152°C
Quantity 25 g
Synonym methyl 3,5-dibromosalicylate, benzoic acid, 3,5-dibromo-2-hydroxy-, methyl ester, 3,5-dibromo-2-hydroxybenzoic acid methyl ester, pubchem3976, acmc-1cpm5, rarechem al bf 0030, timtec-bb sbb001273, 3,5-dibromo-2-hydroxymethyl benzoate, benzoic acid,3,5-dibromo-2-hydroxy-, methyl ester
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-2-hydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.