missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5-hydroxybenzoate, 97%, Thermo Scientific™

Product Code. 10071094
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071094 1 g Amber Glass Bottle 1g
10360814 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071094 Supplier Thermo Scientific Maybridge Supplier No. MO08016DA
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 67973-80-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09263220
InChI Key DMNGQQIFOZYIRA-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxy-benzoic acid methyl ester, benzoic acid, 3-amino-5-hydroxy-, methyl ester, pubchem17212, acmc-1bgay, dmngqqifozyira-uhfffaoysa, methyl 3-amino-5-hydroxy-benzoate, 3-amino-5-methoxycarbonyl phenol, 3-hydroxy-5-methoxycarbonyl aniline, 3-amino-5-hydroxybenzoic acid methyl ester, 5-amino-3-hydroxybenzoic acid methyl ester
PubChem CID 12438277
IUPAC Name methyl 3-amino-5-hydroxybenzoate
SMILES COC(=O)C1=CC(=CC(=C1)O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.