missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyano-5-fluorobenzoate, 98%, Thermo Scientific™

Product Code. p-7030875
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444387 250 mg 250mg
15434387 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444387 Supplier Thermo Scientific Alfa Aesar Supplier No. H31759.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886732-29-2
Molecular Formula C9H6FNO2
Molecular Weight (g/mol) 179.15
MDL Number MFCD08688582
InChI Key YZDWAFGKJVAXAM-UHFFFAOYSA-N
Synonym 3-cyano-5-fluorobenzoic acid methyl ester, benzoic acid, 3-cyano-5-fluoro-, methyl ester
PubChem CID 26597287
IUPAC Name methyl 3-cyano-5-fluorobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)C#N)F

Specifications

Quantity 250 mg
Formula Weight 179.15
Percent Purity 98%
Chemical Name or Material Methyl 3-cyano-5-fluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.