missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trifluoromethyl)phenylacetate, 95%

Product Code. 15435647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435647 1 g 1g
15445647 5 g 5g
15455647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435647 Supplier Thermo Scientific Alfa Aesar Supplier No. H32883.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 62451-84-7 | C10H9F3O2 | 218.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 62451-84-7
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD04039762
InChI Key GMEBDKGPPKAPEM-UHFFFAOYSA-N
PubChem CID 2734603
IUPAC Name methyl 2-[3-(trifluoromethyl)phenyl]acetate
SMILES COC(=O)CC1=CC(=CC=C1)C(F)(F)F

Spécification

Density 1.24
Boiling Point 211°C
Refractive Index 1.45
Quantity 1 g
Synonym methyl 2-3-trifluoromethyl phenyl acetate, methyl 3-trifluoromethyl phenylacetate, methyl 3-trifluoromethylphenyl acetate, methyl 3-trifluoromethyl phenyl acetate, methyltrifluoromethylphenylacetate, methyl m-trifluoromethylphenylacetate, methyl 3-trifluoromethylphenylacetate, methyl-3-trifluoromethylphenylacetate, methyl2-3-trifluoromethyl phenyl acetate
Formula Weight 218.18
Percent Purity 95%
Chemical Name or Material Methyl 3-(trifluoromethyl)phenylacetate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.