missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxyphenylacetate, Mikromol™

Product Code. 30051491
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051491 25 mg 25mg
30052482 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 30051491

Brand: Mikromol MM0013.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-Hydroxyphenylacetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Hydroxyphenylacetate
CAS 14199-15-6
Analyte or Component Names Methyl 4-Hydroxyphenylacetate
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 17025
Synonym 4-Hydroxy-benzeneacetic acid methyl ester, Acetic acid, (p-hydroxyphenyl)-, methyl ester (6CI,7CI,8CI), (4-Hydroxyphenyl)acetic acid methyl ester, 4-Hydroxybenzeneacetic acid methyl ester, Methyl (4-hydroxyphenyl)acetate, Methyl (p-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate, Methyl 2-(p-hydroxyphenyl)acetate, Methyl 4-hydroxybenzeneacetate, p-Hydroxyphenylacetic acid methyl ester, Methyl 4-Hydroxyphenylacetate
InChI Formula InChI=1S/C9H10O3/c1-12-9(11)6-7-2-4-8(10)5-3-7/h2-5,10H,6H2,1H3
SMILES COC(=O)Cc1ccc(O)cc1
IUPAC Name methyl 2-(4-hydroxyphenyl)acetate
Molecular Weight (g/mol) 166.17
Quantity 25 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.