missing translation for 'onlineSavingsMsg'
Learn More

Methyl (5-((2-methyl-4-oxopentan-2-yl)thio)-1H-benzo[d]imidazol-2-yl)carbamate, TRC

Product Code. 30471773
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471773 1 mg 1mg
30476812 5 mg 5mg
30478428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471773 Supplier Toronto Research Chemicals Supplier No. TRCM3057551MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (5-((2-methyl-4-oxopentan-2-yl)thio)-1H-benzo[d]imidazol-2-yl)carbamate
Analyte or Component Names Methyl (5-((2-methyl-4-oxopentan-2-yl)thio)-1H-benzo[d]imidazol-2-yl)carbamate
Shipping Condition Room Temperature
Molecular Formula C15H19N3O3S
Synonym Albendazole Impurity
Solution Type Neat
SMILES COC(=O)Nc1nc2cc(SC(C)(C)CC(=O)C)ccc2[nH]1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 321.39
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.